Methyl-6-{[(4-methylphenyl)sulfonyl]imino}-3(1H)-pyridinecarboxylate ≥95%

Supplier: Matrix Scientific

045156-500MG 045156-1G 045156-5G
101869-412EA 556.92 USD
101869-412 101869-414 101869-416
Methyl-6-{[(4-methylphenyl)sulfonyl]imino}-3(1H)-pyridinecarboxylate ≥95%
Methyl-6-{[(4-methylphenyl)sulfonyl]imino}-3(1H)-pyridinecarboxylate

Hazard: Irritant

Formula: C₁₄H₁₄N₂O₄S
MW: 306.34 g/mol
Melting Pt: 214…216 °C
MDL Number: MFCD06200981
CAS Number: 209971-44-8

Order Now

Specification Test Results

Melting point 214-216°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR