Methyl-6-[(tert-butoxycarbonyl)amino]nicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 6-((tert-butoxycarbonyl)amino)nicotinate

045155-500MG 045155-1G 045155-5G
101869-406EA 556.92 USD
101869-406 101869-408 101869-410
Methyl-6-[(tert-butoxycarbonyl)amino]nicotinate ≥95%
Methyl-6-[(tert-butoxycarbonyl)amino]nicotinate

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₄
MW: 252.27 g/mol
Melting Pt: 166…170 °C
MDL Number: MFCD05663797
CAS Number: 144186-11-8

Order Now

Specification Test Results

Melting point 166-170°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR