1-(4-Morpholino-3-nitrophenyl)-1-ethanone ≥95%

Supplier: Matrix Scientific

045154-500MG 045154-1G 045154-5G
101869-400EA 278.46 USD
101869-400 101869-402 101869-404
1-(4-Morpholino-3-nitrophenyl)-1-ethanone ≥95%
1-(4-Morpholino-3-nitrophenyl)-1-ethanone

Hazard: Irritant

Formula: C₁₂H₁₄N₂O₄
Melting Pt: 79…83 °C
MDL Number: MFCD00047410
CAS Number: 144783-46-0

Order Now

Specification Test Results

Melting point 79-83°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR