2-[4-(2,5-Dimethyl-1H-pyrrol-1-yl)benzyl]malononitrile ≥95%

Supplier: Matrix Scientific

045150-500MG 045150-1G
101869-384EA 556.92 USD
101869-384 101869-386
2-[4-(2,5-Dimethyl-1H-pyrrol-1-yl)benzyl]malononitrile ≥95%
2-[4-(2,5-Dimethyl-1H-pyrrol-1-yl)benzyl]malononitrile

Hazard: Irritant

Formula: C₁₆H₁₅N₃
MW: 249.32 g/mol
Melting Pt: 121…124 °C
MDL Number: MFCD05663795
CAS Number: 771552-11-5

Order Now

Specification Test Results

Melting point 121-124°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR