2-{[4-(2,5-Dimethyl-1H-pyrrol-1-yl)phenyl]methylene}malononitrile ≥95%

Supplier: Matrix Scientific

045149-500MG 045149-1G 045149-5G
101869-378EA 556.92 USD
101869-378 101869-380 101869-382
2-{[4-(2,5-Dimethyl-1H-pyrrol-1-yl)phenyl]methylene}malononitrile ≥95%
2-{[4-(2,5-Dimethyl-1H-pyrrol-1-yl)phenyl]methylene}malononitrile

Hazard: Irritant

Formula: C₁₆H₁₃N₃
Melting Pt: 124…127 °C
MDL Number: MFCD05663794
CAS Number: 771567-64-7

Order Now

Specification Test Results

Melting point 124-127°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR