6-Piperidinonicotinic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
6-(Piperidin-1-yl)nicotinic acid, 6-(1-Piperidinyl)pyridine-3-carboxylic acid, 6-(Piperidin-1-yl)pyridine-3-carboxylic acid
045140-500MG
045140-1G
045140-5G
101869-324EA
173.85
USD
101869-324
101869-326
101869-328
6-Piperidinonicotinic acid ≥95%
6-Piperidinonicotinic acid
Hazard: Irritant
Formula:
C₁₁H₁₄N₂O₂ MW: 206.2411 g/mol Melting Pt: 212-215°C |
MDL Number:
MFCD03791231 CAS Number: 120800-50-2 |
Specification Test Results
Melting point | 212-215°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...