Ethyl-2-anilino-4-oxo-4,5-dihydro-3-furancarboxylate ≥95%

Supplier: Matrix Scientific

045138-500MG 045138-1G 045138-5G
101869-314EA 181.54 USD
101869-314 101869-316 101869-318
Ethyl-2-anilino-4-oxo-4,5-dihydro-3-furancarboxylate ≥95%
Ethyl-2-anilino-4-oxo-4,5-dihydro-3-furancarboxylate

Hazard: Irritant

Formula: C₁₃H₁₃NO₄
Melting Pt: 118…120 °C
MDL Number: MFCD00128484
CAS Number: 58337-16-9

Order Now

Specification Test Results

Melting point 118-120°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR