1-[(4-Chloro-3-nitrophenyl)sulfonyl]-4-piperidinecarboxylic acid ≥95%

Supplier: Matrix Scientific

045136-500MG 045136-1G 045136-5G
101869-304EA 181.54 USD
101869-304 101869-306 101869-308
1-[(4-Chloro-3-nitrophenyl)sulfonyl]-4-piperidinecarboxylic acid ≥95%
1-[(4-Chloro-3-nitrophenyl)sulfonyl]-4-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₃ClN₂O₆S
Melting Pt: 104…142 °C
MDL Number: MFCD03787468
CAS Number: 368427-83-2

Order Now

Specification Test Results

Melting point 104-142°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR