5-Fluoro-2H-3,1-benzoxazine-2,4(1H)-dione ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Fluoro-1H-benzo[d][1,3]oxazine-2,4-dione, 6-Fluoroisatoic anhydride

045134-1G 045134-5G
101869-294EA 430.77 USD
101869-294 101869-296
5-Fluoro-2H-3,1-benzoxazine-2,4(1H)-dione ≥95%
5-Fluoro-2H-3,1-benzoxazine-2,4(1H)-dione

Hazard: Irritant

Formula: C₈H₄FNO₃
MW: 181.12 g/mol
Melting Pt: 230 °C
Density: 1.502
MDL Number: MFCD00112979
CAS Number: 78755-94-9

Order Now

Specification Test Results

Melting point 230°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR