2'-Amino-4'-methoxyacetanilide ≥95%

Supplier: Matrix Scientific

045131-500MG 045131-1G 045131-5G
101869-286EA 130.77 USD
101869-286 101869-288 101869-290
2'-Amino-4'-methoxyacetanilide ≥95%
2'-Amino-4'-methoxyacetanilide

Hazard: Irritant

Formula: C₉H₁₂N₂O₂
MW: 180.2 g/mol
Melting Pt: 150-153°C
MDL Number: MFCD00026124
CAS Number: 5472-37-7

Order Now

Specification Test Results

Melting point 150-153°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR