4-[(3-Formyl-2-pyridinyl)oxy]benzenecarbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 4-[(3-Formylpyridin-2-yl)oxy]benzonitrile

045129-500MG 045129-1G 045129-5G
101869-276EA 375.38 USD
101869-276 101869-278 101869-280
4-[(3-Formyl-2-pyridinyl)oxy]benzenecarbonitrile ≥95%
4-[(3-Formyl-2-pyridinyl)oxy]benzenecarbonitrile

Hazard: Irritant

Formula: C₁₃H₈N₂O₂
MW: 224.22 g/mol
Melting Pt: 162…166 °C
MDL Number: MFCD03305730
CAS Number: 866049-85-6

Order Now

Specification Test Results

Melting point 162-166°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR