Ethyl-4H-thieno[3,2-b]pyrrole-5-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 4H-thieno[3,2-b]pyrrole-5-carboxylate

045127-500MG 045127-1G
101869-268EA 80 USD
101869-268 101869-270
Ethyl-4H-thieno[3,2-b]pyrrole-5-carboxylate ≥95%
Ethyl-4H-thieno[3,2-b]pyrrole-5-carboxylate

Hazard: Irritant

Formula: C₉H₉NO₂S
MW: 195.2383 g/mol
Melting Pt: 131…132 °C
MDL Number: MFCD03425800
CAS Number: 46193-76-4

Order Now

Specification Test Results

Melting point 131-132°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR