11-(Methacryloylamino)undecanoic acid ≥95%
Supplier: Matrix Scientific
045125-500MG
045125-1G
045125-5G
101869-256EA
207.69
USD
101869-256
101869-258
101869-260
11-(Methacryloylamino)undecanoic acid ≥95%
11-(Methacryloylamino)undecanoic acid
Hazard: Irritant
Formula:
C₁₅H₂₇NO₃ Melting Pt: 73…74 °C |
MDL Number:
MFCD03425798 CAS Number: 59178-93-7 |
Specification Test Results
Melting point | 73-74°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...