2-Phenethyl-1-(trifluoromethyl)-1,2,5,6-tetrahydro-4H-pyrrolo[3,2,1-ij]quinolin-1-ol ≥95%

Supplier: Matrix Scientific

045124-500MG 045124-1G
101869-252EA 1483.08 USD
101869-252 101869-254
2-Phenethyl-1-(trifluoromethyl)-1,2,5,6-tetrahydro-4H-pyrrolo[3,2,1-ij]quinolin-1-ol ≥95%
2-Phenethyl-1-(trifluoromethyl)-1,2,5,6-tetrahydro-4H-pyrrolo[3,2,1-ij]quinolin-1-ol

Hazard: Irritant

Formula: C₂₀H₂₀F₃NO
Melting Pt: 76…78 °C
MDL Number: MFCD03425797
CAS Number: 439095-60-0

Order Now

Specification Test Results

Melting point 76-78°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR