1-[3-(Aminocarbonyl)-4-nitrophenyl]-4-piperidinecarboxylic acid ≥95%

Supplier: Matrix Scientific

045122-500MG 045122-1G 045122-5G
101869-242EA 181.54 USD
101869-242 101869-244 101869-246
1-[3-(Aminocarbonyl)-4-nitrophenyl]-4-piperidinecarboxylic acid ≥95%
1-[3-(Aminocarbonyl)-4-nitrophenyl]-4-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₅N₃O₅
Melting Pt: 167…170 °C
MDL Number: MFCD03425795
CAS Number: 439095-54-2

Order Now

Specification Test Results

Melting point 167-170°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR