2,6-Dihydroxy-5-{[4-(4-methoxyphenyl)piperazino]methyl}-4-methylnicotinonitrile ≥95%

Supplier: Matrix Scientific

045117-500MG 045117-1G 045117-5G
101869-222EA 278.46 USD
101869-222 101869-224 101869-226
2,6-Dihydroxy-5-{[4-(4-methoxyphenyl)piperazino]methyl}-4-methylnicotinonitrile ≥95%
2,6-Dihydroxy-5-{[4-(4-methoxyphenyl)piperazino]methyl}-4-methylnicotinonitrile

Hazard: Irritant

Formula: C₁₉H₂₂N₄O₃
Melting Pt: 193…195 °C
MDL Number: MFCD03425792
CAS Number: 439095-50-8

Order Now

Specification Test Results

Melting point 193-195°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR