Ethyl-2-(5-chloro-4-formyl-3-methyl-1H-pyrazol-1-yl)acetate ≥95%

Supplier: Matrix Scientific

045116-500MG 045116-1G
101869-218EA 278.46 USD
101869-218 101869-220
Ethyl-2-(5-chloro-4-formyl-3-methyl-1H-pyrazol-1-yl)acetate ≥95%
Ethyl-2-(5-chloro-4-formyl-3-methyl-1H-pyrazol-1-yl)acetate

Hazard: Irritant

Formula: C₉H₁₁ClN₂O₃
Melting Pt: 66…68 °C
MDL Number: MFCD01837169
CAS Number: 263553-80-6

Order Now

Specification Test Results

Melting point 66-68°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR