2-[(2-Chloro-1,3-thiazol-5-yl)methyl]-1H-isoindole-1,3(2H)-dione ≥95%

Supplier: Matrix Scientific

045113-500MG 045113-1G 045113-5G
101869-206EA 181.54 USD
101869-206 101869-208 101869-210
2-[(2-Chloro-1,3-thiazol-5-yl)methyl]-1H-isoindole-1,3(2H)-dione ≥95%
2-[(2-Chloro-1,3-thiazol-5-yl)methyl]-1H-isoindole-1,3(2H)-dione

Hazard: Irritant

Formula: C₁₂H₇ClN₂O₂S
Melting Pt: 107…108 °C
MDL Number: MFCD03617669
CAS Number: 120740-09-2

Order Now

Specification Test Results

Melting point 107-108°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR