1-(2-fluoro-4-nitrophenyl)-4-methylpiperazine ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(2-Fluoro-4-nitrophenyl)-4-methylpiperazine

045111-500MG 045111-1G 045111-5G
101869-194EA 181.54 USD
101869-194 101869-196 101869-198
1-(2-fluoro-4-nitrophenyl)-4-methylpiperazine ≥95%
1-(2-fluoro-4-nitrophenyl)-4-methylpiperazine

Hazard: Irritant

Formula: C₁₁H₁₄FN₃O₂
MW: 239.25 g/mol
Melting Pt: 71…72 °C
MDL Number: MFCD02633484
CAS Number: 221198-29-4

Order Now

Specification Test Results

Melting point 71-72°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR