3-Amino-3-[3-(trifluoromethyl)phenyl]propanoic acid ≥95%

Supplier: Matrix Scientific

045107-500MG 045107-1G
101869-184EA 556.92 USD
101869-184 101869-186
3-Amino-3-[3-(trifluoromethyl)phenyl]propanoic acid ≥95%
3-Amino-3-[3-(trifluoromethyl)phenyl]propanoic acid

Hazard: Irritant

Formula: C10H10F3NO2
MW: 233.1900 g/mol
Melting Pt: 210…212 °C
MDL Number: MFCD02653329
CAS Number: 143438-91-9

Order Now

Specification Test Results

Melting point 210-212°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR