4-Phenyl-1H-pyrazolo[3,4-b]pyridin-3-ylamine ≥95%

Supplier: Matrix Scientific

045097-500MG 045097-1G
101869-162EA 556.92 USD
101869-162 101869-164
4-Phenyl-1H-pyrazolo[3,4-b]pyridin-3-ylamine ≥95%
4-Phenyl-1H-pyrazolo[3,4-b]pyridin-3-ylamine

Hazard: Irritant

Formula: C₁₂H₁₀N₄
Melting Pt: 262…264 °C
MDL Number: MFCD03001275
CAS Number: 174713-63-4

Order Now

Specification Test Results

Melting point 262-264°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR