1-Chloro-3-iodo-2-nitrobenzene ≥95%

Supplier: Matrix Scientific

045088-500MG 045088-1G 045088-5G
101869-114EA 41.54 USD
101869-114 101869-116 101869-118
1-Chloro-3-iodo-2-nitrobenzene ≥95%
1-Chloro-3-iodo-2-nitrobenzene

Hazard: Irritant

Formula: C₆H₃ClINO₂
MW: 283.4510 g/mol
Melting Pt: 60-61°C
MDL Number: MFCD08443947
CAS Number: 937601-42-8

Order Now

Specification Test Results

Melting point 60-61°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR