1-[2-(4-Chlorophenyl)-4-methyl-1,3-thiazol-5-yl]-1-ethanone ≥95%

Supplier: Matrix Scientific

045086-500MG 045086-1G 045086-5G
101869-102EA 278.46 USD
101869-102 101869-104 101869-106
1-[2-(4-Chlorophenyl)-4-methyl-1,3-thiazol-5-yl]-1-ethanone ≥95%
1-[2-(4-Chlorophenyl)-4-methyl-1,3-thiazol-5-yl]-1-ethanone

Hazard: Irritant

Formula: C₁₂H₁₀ClNOS
Melting Pt: 125…127 °C
MDL Number: MFCD01871615
CAS Number: 54001-07-9

Order Now

Specification Test Results

Melting point 125-127°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR