1-Bromo-4-chloro-3-nitrobenzene ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Bromo-1-chloro-2-nitrobenzene, 5-Bromo-2-chloronitrobenzene

045084-5G 045084-25G
101869-094EA 32.31 USD
101869-094 102551-338
1-Bromo-4-chloro-3-nitrobenzene ≥95%
1-Bromo-4-chloro-3-nitrobenzene

Hazard: Irritant

Formula: C₆H₃BrClNO₂
MW: 236.4500 g/mol
Melting Pt: 70-72°C
MDL Number: MFCD05863216
CAS Number: 16588-24-2

Order Now

Specification Test Results

Melting point 70-72°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR