4-(4-Chloro-3-fluorophenoxy)benzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

045081-500MG 045081-1G 045081-5G
101869-080EA 438.46 USD
101869-080 101869-082 101869-084
4-(4-Chloro-3-fluorophenoxy)benzenecarboxylic acid ≥95%
4-(4-Chloro-3-fluorophenoxy)benzenecarboxylic acid

Hazard: Irritant

Formula: C₁₃H₈ClFO₃
Melting Pt: 184…186 °C
MDL Number: MFCD01871572
CAS Number: 551921-67-6

Order Now

Specification Test Results

Melting point 184-186°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR