[5-Phenyl-3-isoxazolyl]methanol ≥95%
Supplier: Matrix Scientific
Synonyms:
(5-Phenylisoxazol-3-yl)methanol, 3-Hydroxymethyl-5-phenylisoxazole
045078-500MG
045078-1G
045078-5G
101869-062EA
223.08
USD
101869-062
101869-064
101869-066
[5-Phenyl-3-isoxazolyl]methanol ≥95%
[5-Phenyl-3-isoxazolyl]methanol
Hazard: Irritant
Formula:
C₁₀H₉NO₂ MW: 175.1840 g/mol Melting Pt: 98…100 °C |
MDL Number:
MFCD01444104 CAS Number: 1619-37-0 |
Specification Test Results
Melting point | 98-100°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...