4-(6-Chloro-2-phenyl-4-pyrimidinyl)morpholine ≥95%

Supplier: Matrix Scientific

045077-500MG 045077-1G 045077-5G
101869-056EA 375.38 USD
101869-056 101869-058 101869-060
4-(6-Chloro-2-phenyl-4-pyrimidinyl)morpholine ≥95%
4-(6-Chloro-2-phenyl-4-pyrimidinyl)morpholine

Hazard: Irritant

Formula: C₁₄H₁₄ClN₃O
MW: 275.74 g/mol
Melting Pt: 128…130 °C
MDL Number: MFCD00754198
CAS Number: 343373-72-8

Order Now

Specification Test Results

Melting point 128-130°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR