2-Chloro-N-(5-iodo-2-pyridinyl)acetamide ≥95%

Supplier: Matrix Scientific

045074-500MG 045074-1G 045074-5G
101869-040EA 124.62 USD
101869-040 101869-042 101869-044
2-Chloro-N-(5-iodo-2-pyridinyl)acetamide ≥95%
2-Chloro-N-(5-iodo-2-pyridinyl)acetamide

Hazard: Irritant

Formula: C₇H₆ClIN₂O
MW: 296.5 g/mol
Melting Pt: 175…177 °C
MDL Number: MFCD01315229
CAS Number: 343373-23-9

Order Now

Specification Test Results

Melting point 175-177°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR