4-Bromo-N-(2-hydroxyethyl)benzenecarboxamide ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Bromo-N-(2-hydroxyethyl)benzamide
045072-500MG
045072-1G
045072-5G
101869-028EA
181.54
USD
101869-028
101869-030
101869-032
4-Bromo-N-(2-hydroxyethyl)benzenecarboxamide ≥95%
4-Bromo-N-(2-hydroxyethyl)benzenecarboxamide
Hazard: Irritant
Formula:
C₉H₁₀BrNO₂ MW: 244.09 g/mol Melting Pt: 146…147 °C |
MDL Number:
MFCD00456170 CAS Number: 57728-67-3 |
Specification Test Results
Melting point | 146-147°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...