(3-Phenyl-5-isoxazolyl)methanamine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-(Aminomethyl)-3-phenylisoxazole, (3-Phenylisoxazol-5-yl)methanamine

045071-500MG 045071-1G 045071-5G
101869-022EA 361.54 USD
101869-022 101869-024 101869-026
(3-Phenyl-5-isoxazolyl)methanamine ≥95%
(3-Phenyl-5-isoxazolyl)methanamine

Hazard: Irritant

Formula: C₁₀H₁₀N₂O
MW: 174.2 g/mol
Melting Pt: 51…53 °C
MDL Number: MFCD03844852
CAS Number: 54408-35-4

Order Now

Specification Test Results

Melting point 51-53°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR