[3-(4-Methoxyphenyl)-5-isoxazolyl]methanamine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-(Aminomethyl)-3-(4-methoxyphenyl)isoxazole

045070-500MG 045070-1G
101869-018EA 310.77 USD
101869-018 101869-020
[3-(4-Methoxyphenyl)-5-isoxazolyl]methanamine ≥95%
[3-(4-Methoxyphenyl)-5-isoxazolyl]methanamine

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₂
MW: 204.23 g/mol
Melting Pt: 119…122 °C
MDL Number: MFCD01935989
CAS Number: 338982-43-7

Order Now

Specification Test Results

Melting point 119-122°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR