4-[(5-Formyl-2-pyridinyl)oxy]benzenecarbonitrile ≥95%

Supplier: Matrix Scientific

045067-500MG 045067-1G 045067-5G
101869-002EA 556.92 USD
101869-002 101869-004 101869-006
4-[(5-Formyl-2-pyridinyl)oxy]benzenecarbonitrile ≥95%
4-[(5-Formyl-2-pyridinyl)oxy]benzenecarbonitrile

Hazard: Irritant

Formula: C₁₃H₈N₂O₂
Melting Pt: 133…135.5 °C
MDL Number: MFCD00974909
CAS Number: 328547-41-7

Order Now

Specification Test Results

Melting point 133-135.5°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR