5-Chloro-3-phenyl-1H-indole-2-carbonitrile ≥95%
Supplier: Matrix Scientific
045065-500MG
045065-1G
045065-5G
101868-990EA
1483.08
USD
101868-990
101868-992
101868-994
5-Chloro-3-phenyl-1H-indole-2-carbonitrile ≥95%
5-Chloro-3-phenyl-1H-indole-2-carbonitrile
Hazard: Irritant
Formula:
C₁₅H₉ClN₂ |
MDL Number:
MFCD00974247 CAS Number: 24139-17-1 |
Specification Test Results
Melting point | 213-215°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...