5-Chloro-3-phenyl-1H-indole-2-carboxylic acid ≥95%
Supplier: Matrix Scientific
045063-500MG
045063-1G
045063-5G
101868-982EA
921.54
USD
101868-982
101868-984
101868-986
5-Chloro-3-phenyl-1H-indole-2-carboxylic acid ≥95%
5-Chloro-3-phenyl-1H-indole-2-carboxylic acid
Hazard: Irritant
Formula:
C₁₅H₁₀ClNO₂ Melting Pt: 230…232 °C |
MDL Number:
MFCD00425492 CAS Number: 21139-31-1 |
Specification Test Results
Melting point | 230-232°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...