Ethyl-5-chloro-3-phenyl-1H-indole-2-carboxylate ≥95%

Supplier: Matrix Scientific

045062-500MG 045062-1G 045062-5G
101868-976EA 747.69 USD
101868-976 101868-978 101868-980
Ethyl-5-chloro-3-phenyl-1H-indole-2-carboxylate ≥95%
Ethyl-5-chloro-3-phenyl-1H-indole-2-carboxylate

Hazard: Irritant

Formula: C₁₇H₁₄ClNO₂
Melting Pt: 176…178 °C
MDL Number: MFCD00411467
CAS Number: 21139-32-2

Order Now

Specification Test Results

Melting point 176-178°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR