Diethyl-2-oxo-1H-1,3-benzimidazole-1,3(2H)-dicarboxylate ≥95%

Supplier: Matrix Scientific

045061-500MG 045061-1G 045061-5G
101868-970EA 181.54 USD
101868-970 101868-972 101868-974
Diethyl-2-oxo-1H-1,3-benzimidazole-1,3(2H)-dicarboxylate ≥95%
Diethyl-2-oxo-1H-1,3-benzimidazole-1,3(2H)-dicarboxylate

Hazard: Irritant

Formula: C₁₃H₁₄N₂O₅
Melting Pt: 123…126 °C
MDL Number: MFCD08056645
CAS Number: 161468-57-1

Order Now

Specification Test Results

Melting point 123-126°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR