2-(Methylsulfanyl)benzenecarbohydrazide ≥95%

Supplier: Matrix Scientific

045060-500MG 045060-1G 045060-5G
101868-964EA 375.38 USD
101868-964 101868-966 101868-968
2-(Methylsulfanyl)benzenecarbohydrazide ≥95%
2-(Methylsulfanyl)benzenecarbohydrazide

Hazard: Irritant

Formula: C₈H₁₀N₂OS
Melting Pt: 102…104 °C
MDL Number: MFCD06200979
CAS Number: 885949-42-8

Order Now

Specification Test Results

Melting point 102-104°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR