4-Bromophenyl-4-(chloromethyl)-1-methyl-3-phenyl-1H-pyrazol-5-yl sulfide ≥95%

Supplier: Matrix Scientific

045059-500MG 045059-1G 045059-5G
101868-958EA 921.54 USD
101868-958 101868-960 101868-962
4-Bromophenyl-4-(chloromethyl)-1-methyl-3-phenyl-1H-pyrazol-5-yl sulfide ≥95%
4-Bromophenyl-4-(chloromethyl)-1-methyl-3-phenyl-1H-pyrazol-5-yl sulfide

Hazard: Irritant

Formula: C₁₇H₁₄BrClN₂S
MW: 393.74 g/mol
Melting Pt: 114…116 °C
MDL Number: MFCD01315357
CAS Number: 318959-24-9

Order Now

Specification Test Results

Melting point 114-116°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR