4-Chlorobenzyl-4-(chloromethyl)-1-methyl-3-phenyl-1H-pyrazol-5-yl sulfide ≥95%

Supplier: Matrix Scientific

045058-500MG 045058-1G 045058-5G
101868-952EA 921.54 USD
101868-952 101868-954 101868-956
4-Chlorobenzyl-4-(chloromethyl)-1-methyl-3-phenyl-1H-pyrazol-5-yl sulfide ≥95%
4-Chlorobenzyl-4-(chloromethyl)-1-methyl-3-phenyl-1H-pyrazol-5-yl sulfide

Hazard: Irritant

Formula: C₁₈H₁₆Cl₂N₂S
MW: 363.31 g/mol
Melting Pt: 71…73 °C
MDL Number: MFCD01315356
CAS Number: 318959-23-8

Order Now

Specification Test Results

Melting point 71-73°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR