4-(Chloromethyl)-5-(2,4-dichlorophenoxy)-1-methyl-3-(trifluoromethyl)-1H-pyrazole ≥95%

Supplier: Matrix Scientific

045057-500MG 045057-1G 045057-5G
101868-946EA 921.54 USD
101868-946 101868-948 101868-950
4-(Chloromethyl)-5-(2,4-dichlorophenoxy)-1-methyl-3-(trifluoromethyl)-1H-pyrazole ≥95%
4-(Chloromethyl)-5-(2,4-dichlorophenoxy)-1-methyl-3-(trifluoromethyl)-1H-pyrazole

Hazard: Irritant

Formula: C₁₂H₈Cl₃F₃N₂O
Melting Pt: 54…56 °C
MDL Number: MFCD00664860
CAS Number: 318959-15-8

Order Now

Specification Test Results

Melting point 54-56°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR