[5-(2,4-Dichlorophenoxy)-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol ≥95%
Supplier: Matrix Scientific
045056-500MG
045056-1G
045056-5G
101868-940EA
921.54
USD
101868-940
101868-942
101868-944
[5-(2,4-Dichlorophenoxy)-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol ≥95%
[5-(2,4-Dichlorophenoxy)-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol
Hazard: Irritant
Formula:
C₁₂H₉Cl₂F₃N₂O₂ Melting Pt: 79…83 °C |
MDL Number:
MFCD06203030 CAS Number: 1006491-86-6 |
Specification Test Results
Melting point | 79-83°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...