3-Hydroxy-2-methyl-4(1H)-pyridinone ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Hydroxy-2-methylpyridin-4(1H)-one

045054-500MG 045054-1G 045054-5G
101868-928EA 230.77 USD
101868-928 101868-930 101868-932
3-Hydroxy-2-methyl-4(1H)-pyridinone ≥95%
3-Hydroxy-2-methyl-4(1H)-pyridinone

Hazard: Irritant

Formula: C₆H₇NO₂
MW: 125.13 g/mol
Melting Pt: 300 °C
MDL Number: MFCD02585559
CAS Number: 17184-19-9

Order Now

Specification Test Results

Melting point 300°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR