3-[(4-Fluorobenzyl)oxy]-2-methyl-4(1H)-pyridinone 95

Supplier: Matrix Scientific

045053-500MG 045053-1G
101868-924EA 264.62 USD
101868-924 101868-926
3-[(4-Fluorobenzyl)oxy]-2-methyl-4(1H)-pyridinone 95
3-[(4-Fluorobenzyl)oxy]-2-methyl-4(1H)-pyridinone

Hazard: Irritant

Formula: C₁₃H₁₂FNO₂
MW: 233.24 g/mol
Melting Pt: 129…134 °C
MDL Number: MFCD00215663
CAS Number: 338965-51-8

Order Now

Specification Test Results

Melting point 129-134°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR