2-[(4-Nitrophenyl)methylene]malononitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(4-Nitrobenzylidene)malononitrile

045048-500MG 045048-1G 045048-5G
101868-898EA 181.54 USD
101868-898 101868-900 101868-902
2-[(4-Nitrophenyl)methylene]malononitrile ≥95%
2-[(4-Nitrophenyl)methylene]malononitrile

Hazard: Irritant

Formula: C₁₀H₅N₃O₂
MW: 199.17 g/mol
Melting Pt: 149…151 °C
MDL Number: MFCD00159003
CAS Number: 2700-23-4

Order Now

Specification Test Results

Melting point 149-151°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR