N-(2-Hydroxy-1,1-dimethylethyl)-2-methoxybenzenecarboxamide ≥95%

Supplier: Matrix Scientific

Synonyms: N-(1-Hydroxy-2-methylpropan-2-yl)-2-methoxybenzamide

045043-500MG 045043-1G 045043-5G
101868-868EA 278.46 USD
101868-868 101868-870 101868-872
N-(2-Hydroxy-1,1-dimethylethyl)-2-methoxybenzenecarboxamide ≥95%
N-(2-Hydroxy-1,1-dimethylethyl)-2-methoxybenzenecarboxamide

Hazard: Irritant

Formula: C₁₂H₁₇NO₃
MW: 223.27 g/mol
Melting Pt: 129…131 °C
MDL Number: MFCD00665208
CAS Number: 74201-13-1

Order Now

Specification Test Results

Melting point 129-131°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR