Ethyl-1-[[3-chloro-5-(trifluoromethyl)-2-pyridinyl](methyl)amino]-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarboxyl ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 1-[[3-chloro-5-(trifluoromethyl)pyridin-2-yl](methyl)amino]-2-methyl-6-oxo-1,4,5,6-tetrahydrop

045042-500MG 045042-1G
101868-864EA 1109.23 USD
101868-864 101868-866
Ethyl-1-[[3-chloro-5-(trifluoromethyl)-2-pyridinyl](methyl)amino]-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarboxyl ≥95%
Ethyl-1-[[3-chloro-5-(trifluoromethyl)-2-pyridinyl](methyl)amino]-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarboxyl

Hazard: Irritant

Formula: C₁₆H₁₇ClF₃N₃O₃
MW: 391.78 g/mol
Melting Pt: 75…77 °C
MDL Number: MFCD01568671
CAS Number: 338965-01-8

Order Now

Specification Test Results

Melting point 75-77°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR