2-Oxo-5-[3-(trifluoromethyl)phenyl]-1,2-dihydro-3-pyridinecarbonitrile ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Cyano-5-[(trifluoromethyl)phenyl]-2(1H)-pyridone
045041-500MG
045041-1G
045041-5G
101868-858EA
747.69
USD
101868-858
101868-860
101868-862
2-Oxo-5-[3-(trifluoromethyl)phenyl]-1,2-dihydro-3-pyridinecarbonitrile ≥95%
2-Oxo-5-[3-(trifluoromethyl)phenyl]-1,2-dihydro-3-pyridinecarbonitrile
Hazard: Irritant
Formula:
C₁₃H₇F₃N₂O MW: 264.21 g/mol Melting Pt: 234…237 °C |
MDL Number:
MFCD00232370 CAS Number: 76053-36-6 |
Specification Test Results
Melting point | 234-237°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...