2-Chloro-5-(3-chlorophenyl)nicotinonitrile ≥95%

Supplier: Matrix Scientific

045040-500MG 045040-1G 045040-5G
101868-852EA 747.69 USD
101868-852 101868-854 101868-856
2-Chloro-5-(3-chlorophenyl)nicotinonitrile ≥95%
2-Chloro-5-(3-chlorophenyl)nicotinonitrile

Hazard: Irritant

Formula: C₁₂H₆Cl₂N₂
Melting Pt: 173…176 °C
MDL Number: MFCD00231620
CAS Number: 338964-13-9

Order Now

Specification Test Results

Melting point 173-176°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR