Ethyl-8-cyano-4-hydroxyquinoline-3-carboxylate ≥95%
Supplier: Matrix Scientific
045038-500MG
045038-1G
045038-5G
101868-840EA
556.92
USD
101868-840
101868-842
101868-844
Ethyl-8-cyano-4-hydroxyquinoline-3-carboxylate ≥95%
Ethyl 8-cyano-4-hydroxyquinoline-3-carboxylate
Hazard: Irritant
Formula:
C₁₃H₁₀N₂O₃ Melting Pt: 235…238 °C |
MDL Number:
MFCD00173411 CAS Number: 77156-79-7 |
Specification Test Results
Melting point | 235-238°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...