Ethyl-4-chloroquinoline-3-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Chloroquinoline-3-carboxylic acid ethyl ester, Ethyl 4-chloroquinoline-3-carboxylate
045037-500MG
045037-1G
045037-5G
101868-834EA
556.92
USD
101868-834
101868-836
101868-838
Ethyl-4-chloroquinoline-3-carboxylate ≥95%
Ethyl 4-chloroquinoline-3-carboxylate
Hazard: Irritant
Formula:
C₁₂H₁₀ClNO₂ MW: 235.67 g/mol Melting Pt: 46-48°C |
MDL Number:
MFCD00173407 CAS Number: 13720-94-0 |
Specification Test Results
Melting point | 46-48°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...